3-(Diisopropylcarbamoyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR4096-500MG
APOSOR4096-500MGEA
21.9
EUR
APOSOR4096-500MG
3-(Diisopropylcarbamoyl)phenylboronsäure 98%
3-(Diisopropylcarbamoyl)phenylboronsäure
Formel:
C₁₃H₂₀BNO₃ Schmelzpunkt: 170…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659890 CAS-Nummer: 850567-40-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...