N-(2-Hydroxyethyl)imidazo[1,2-a]pyridine-2-carboxamide
Lieferant: Apollo Scientific
OR41008-1G
APOSOR41008-1GEA
342
EUR
APOSOR41008-1G
N-(2-Hydroxyethyl)imidazo[1,2-a]pyridine-2-carboxamide
N-(2-Hydroxyethyl)imidazo[1,2-a]pyridine-2-carboxamide
Formel:
C₁₀H₁₁N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12179438 CAS-Nummer: 1157107-85-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...