3-Methylmorpholine 97%

Lieferant: Apollo Scientific
Achtung

OR41010-5G
APOSOR41010-5GEA 58 EUR
APOSOR41010-5G
3-Methylmorpholine 97%
3-Methylmorpholine
Formel: C5H11NO
Lagertemperatur: Kühlschrank
MDL: MFCD09054773
CAS-Nummer: 42185-06-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR