(2,3-Dihydro-3-oxo-4H-1,4-benzoxazin-4-yl)acetonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Cyanomethyl)-3,4-dihydro-3-oxo-2H-1,4-benzoxazine, (2,3-Dihydro-3-oxo-4H-benzo[b][1,4]oxazin-4-yl)acetonitrile

OR4102-1G
APOSOR4102-1GEA 667 EUR
APOSOR4102-1G
(2,3-Dihydro-3-oxo-4H-1,4-benzoxazin-4-yl)acetonitrile
(2,3-Dihydro-3-oxo-4H-1,4-benzoxazin-4-yl)acetonitrile
Formel: C₁₀H₈N₂O₂
Molecular Weight: 188,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00268751
CAS-Nummer: 115615-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR