2,3,4,4'-Tetrahydroxybenzophenon 98%

Lieferant: Apollo Scientific

Synonyme: (4-Hydroxyphenyl)(2,3,4-trihydroxyphenyl)methanone

OR41023-100G
APOSOR41023-100GEA 122 EUR
APOSOR41023-100G
2,3,4,4'-Tetrahydroxybenzophenon 98%
2,3,4,4'-Tetrahydroxybenzophenon
Formel: C₁₃H₁₀O₅
Molecular Weight: 246.2155 g/mol
Schmelzpunkt: 221 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00083097
CAS-Nummer: 31127-54-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR