1-Adamantyl methacrylate
Lieferant: Apollo Scientific
|
Achtung
|
OR41031-100G
OR41031-25G
OR41031-5G
APOSOR41031-100GEA
0
EUR
APOSOR41031-100G
APOSOR41031-25G
APOSOR41031-5G
1-Adamantyl methacrylate
1-Adamantyl methacrylate
Formel:
C₁₄H₂₀O₂ Siedepunkt: 113 °C/3mmHg Dichte: 1,0495 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD08703330 CAS-Nummer: 16887-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...