Ethyl-6-bromoindole-2-carboxylate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 6-bromoindole-2-carboxylate
OR41055-5G
APOSOR41055-5GEA
109
EUR
APOSOR41055-5G
Ethyl-6-bromoindole-2-carboxylate 98%
Ethyl-6-bromoindole-2-carboxylate
Formel:
C₁₁H₁₀BrNO₂ Molecular Weight: 268.11 g/mol Schmelzpunkt: 178…188 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03094942 CAS-Nummer: 103858-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...