4-Methoxy-2,5-dimethylphenylboronic acid 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Borono-2,5-dimethylanisole, 4-Methoxy-2,5-dimethylbenzeneboronic acid, (4-Methoxy-2,5-dimethylphenyl)boronic acid, 2,5-Dimethyl-4-methoxyphenylboronic acid, 2,5-Dimethyl-4-methoxybenzeneboronic acid
OR4108-250MG
APOSOR4108-250MGEA
21.9
EUR
APOSOR4108-250MG
4-Methoxy-2,5-dimethylphenylboronic acid 96%
4-Methoxy-2,5-dimethylphenylboronic acid
Formel:
C₉H₁₃BO₃ Molecular Weight: 180,01 g/mol Schmelzpunkt: 187 - 190 °C |
MDL:
MFCD06801676 CAS-Nummer: 246023-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...