Methyl 4-azaindole-3-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 1H-pyrrolo[3,2-b]pyridine-3-carboxylate, 3-(Methoxycarbonyl)-1H-pyrrolo[3,2-b]pyridine

OR41124-50MG
APOSOR41124-50MGEA 21.9 EUR
APOSOR41124-50MG
Methyl 4-azaindole-3-carboxylate 97%
Methyl 4-azaindole-3-carboxylate
Formel: C₉H₈N₂O₂
Molecular Weight: 176,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864655
CAS-Nummer: 952800-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR