Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR41128-25G
APOSOR41128-25GEA 171 EUR
APOSOR41128-25G
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid 97%
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
Formel: C₇H₅N₃O₂
Molecular Weight: 163.136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01975194
CAS-Nummer: 25940-35-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR