1-n-Propyl-4-cyclopropylaminopiperidine 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Cyclopropyl-1-propylpiperidin-4-amine

OR4114-250MG
APOSOR4114-250MGEA 140 EUR
APOSOR4114-250MG
1-n-Propyl-4-cyclopropylaminopiperidine 95+%
1-n-Propyl-4-cyclopropylaminopiperidine
Formel: C₁₁H₂₂N₂
Molecular Weight: 182,31 g/mol
Siedepunkt: 80 °C/2mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01941332
CAS-Nummer: 387358-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR