2,6-Dimethylbenzoesäure
Lieferant: Apollo Scientific
|
Achtung
|
OR41144-25G
APOSOR41144-25GEA
69.6
EUR
APOSOR41144-25G
2,6-Dimethylbenzoesäure
2,6-Dimethylbenzoic acid
Formel:
C₉H₁₀O₂ Molecular Weight: 150.18 g/mol Siedepunkt: 160 °C/17mmHg Schmelzpunkt: 114…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002483 CAS-Nummer: 632-46-2 EINECS: 211-177-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...