3-Bromopiperidine-2,6-dione 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-2,6-dioxopiperidine

OR41150-5G
APOSOR41150-5GEA 71.2 EUR
APOSOR41150-5G
3-Bromopiperidine-2,6-dione 95%
3-Bromopiperidine-2,6-dione
Formel: C₅H₆BrNO₂
Molecular Weight: 192.011 g/mol
Schmelzpunkt: 107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11053059
CAS-Nummer: 62595-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR