3-Iodopyridin-4-ol 97%

Lieferant: Apollo Scientific

OR41171-1G
APOSOR41171-1GEA 0 EUR
APOSOR41171-1G
3-Iodopyridin-4-ol 97%
3-Iodopyridin-4-ol
Formel: C₅H₄INO
Lagertemperatur: Raumtemperatur
MDL: MFCD01646059
CAS-Nummer: 89282-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR