4-(tert-Butoxycarbonyl)phenylboronsäurepinakolester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 4-(tert-Butoxycarbonyl)phenylboronic acid, pinacol ester, 4-Boc-phenylboronic acid Pinacol ester, 2-(4-tert-Butoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid tert-Butyl ester, 4-(tert-Butoxycarbonyl)phenylboronic acid Pinacol ester

OR4118-5G
APOSOR4118-5GEA 0 EUR
APOSOR4118-5G
4-(tert-Butoxycarbonyl)phenylboronsäurepinakolester 97%
4-(tert-Butoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₅BO₄
Molecular Weight: 304,1890 g/mol
Schmelzpunkt: 128…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03453055
CAS-Nummer: 850568-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR