5-Brom-2-methoxy-4-methyl-3-nitropyridin

Lieferant: Apollo Scientific
Achtung

OR41181-1G OR41181-5G
APOSOR41181-1GEA 21.9 EUR
APOSOR41181-1G APOSOR41181-5G
5-Brom-2-methoxy-4-methyl-3-nitropyridin
5-Brom-2-methoxy-4-methyl-3-nitropyridin
Formel: C₇H₇BrN₂O₃
Molecular Weight: 247.0500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659504
CAS-Nummer: 884495-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR