2-(2,3-Dimethylphenyl)acetonitrile

Lieferant: Apollo Scientific
Achtung

OR41183-250MG
APOSOR41183-250MGEA 219 EUR
APOSOR41183-250MG
2-(2,3-Dimethylphenyl)acetonitrile
2-(2,3-Dimethylphenyl)acetonitrile
Formel: C₁₀H₁₁N
Molecular Weight: 145,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09834792
CAS-Nummer: 76574-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR