4-(2-Methoxyethylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(2-Methoxyethylcarbamoyl)benzeneboronic acid, 4-(2-Methoxyethylaminocarbonyl)phenylboronic acid

OR4125-1G
APOSOR4125-1GEA 194 EUR
APOSOR4125-1G
4-(2-Methoxyethylcarbamoyl)phenylboronsäure 98%
4-(2-Methoxyethylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₄BNO₄
Molecular Weight: 223,04 g/mol
Schmelzpunkt: 120…124 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115682
CAS-Nummer: 850589-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR