N-Isopropyl-4-boronobenzolsulfonamid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-(N-Isopropylsulphamoyl)phenyl)boronic acid, 4-(Isopropylsulphamoyl)benzeneboronic acid
OR4127-500MG
APOSOR4127-500MGEA
21.9
EUR
APOSOR4127-500MG
N-Isopropyl-4-boronobenzolsulfonamid 98%
N-Isopropyl-4-boronobenzolsulfonamid
Formel:
C₉H₁₄BNO₄S Molecular Weight: 243,08 g/mol Schmelzpunkt: 136…140 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07363752 CAS-Nummer: 850589-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...