Methyl nicotinoylacetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-oxo-3-(pyridin-3-yl)propanoate

OR4128-1G OR4128-5G OR4128-25G
APOSOR4128-1GEA 29 EUR
APOSOR4128-1G APOSOR4128-5G APOSOR4128-25G
Methyl nicotinoylacetate
Methyl nicotinoylacetate
Formel: C₉H₉NO₃
Molecular Weight: 179.175 g/mol
Schmelzpunkt: 34…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216524
CAS-Nummer: 54950-20-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR