N-Ethyl-4-boronobenzolsulfonamid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Ethylsulphamoyl)benzeneboronic acid

OR4132-1G
APOSOR4132-1GEA 244 EUR
APOSOR4132-1G
N-Ethyl-4-boronobenzolsulfonamid 97%
N-Ethyl-4-boronobenzolsulfonamid
Formel: C₈H₁₂BNO₄S
Molecular Weight: 229,06 g/mol
Schmelzpunkt: 132…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07363751
CAS-Nummer: 871329-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR