5-Hydroxymethyluracil
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Hydroxymethyl)pyrimidine-2,4(1H,3H)-dione
OR4151-25G
OR4151-5G
OR4151-100G
APOSOR4151-25GEA
102
EUR
APOSOR4151-25G
APOSOR4151-5G
APOSOR4151-100G
5-Hydroxymethyluracil
5-Hydroxymethyluracil
Formel:
C₅H₆N₂O₃ Molecular Weight: 142.114 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006070 CAS-Nummer: 4433-40-3 EINECS: 224-636-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...