1-(2-Hydroxyphenyl)piperazin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
o-(1-Piperazinyl)phenol
OR4153-10G
OR4153-250G
OR4153-50G
APOSOR4153-10GEA
81.2
EUR
APOSOR4153-10G
APOSOR4153-250G
APOSOR4153-50G
1-(2-Hydroxyphenyl)piperazin 98%
1-(2-Hydroxyphenyl)piperazin
Formel:
C₁₀H₁₄N₂O Molecular Weight: 178.23 g/mol Siedepunkt: 318 Schmelzpunkt: 125…129 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190246 CAS-Nummer: 1011-17-2 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...