Benzyl-tert-butyl malonate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Benzyl tert-butyl malonate
OR4154-5G
APOSOR4154-5GEA
21.9
EUR
APOSOR4154-5G
Benzyl-tert-butyl malonate 95%
Benzyl-tert-butyl malonate
Formel:
C₁₄H₁₈O₄ Molecular Weight: 250,294 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075175 CAS-Nummer: 72594-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...