1-(3-Methoxypropyl)piperazin

Lieferant: Apollo Scientific
Achtung

OR4159-1G
APOSOR4159-1GEA 46.4 EUR
APOSOR4159-1G
1-(3-Methoxypropyl)piperazin
1-(3-Methoxypropyl)piperazin
Formel: C₈H₁₈N₂O
Siedepunkt: 87 °C/4,5mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD02093537
CAS-Nummer: 88708-40-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR