1-(1-Naphthylmethyl)piperazine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR4166-5G
APOSOR4166-5GEA
0
EUR
APOSOR4166-5G
1-(1-Naphthylmethyl)piperazine 97%
1-(1-Naphthylmethyl)piperazine
Formel:
C₁₅H₁₈N₂ Schmelzpunkt: 63…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01314185 CAS-Nummer: 40675-81-8 UN: 3077 ADR: 9+EHS,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...