1-(1-Ethylpropyl)piperazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Pent-3-yl)piperazine

OR4170-1G
APOSOR4170-1GEA 58 EUR
APOSOR4170-1G
1-(1-Ethylpropyl)piperazine
1-(1-Ethylpropyl)piperazine
Formel: C₉H₂₀N₂
Molecular Weight: 156.27 g/mol
Siedepunkt: 86 °C/11.2mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD02093554
CAS-Nummer: 373356-51-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR