2-(4-Nitrophenoxy)acetohydrazide 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Nitrophenoxyacetic acid hydrazide

OR4176-10G
APOSOR4176-10GEA 36.4 EUR
APOSOR4176-10G
2-(4-Nitrophenoxy)acetohydrazide 98%
2-(4-Nitrophenoxy)acetohydrazide
Formel: C₈H₉N₃O₄
Molecular Weight: 211,18 g/mol
Schmelzpunkt: 184…186 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051973
CAS-Nummer: 75129-74-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR