6-Methyl[1,2,4]triazolo[4,3-b]pyridazine

Lieferant: Apollo Scientific
Achtung

OR42002-250MG
APOSOR42002-250MGEA 63.8 EUR
APOSOR42002-250MG
6-Methyl[1,2,4]triazolo[4,3-b]pyridazine
6-Methyl[1,2,4]triazolo[4,3-b]pyridazine
Formel: C₆H₆N₄
Molecular Weight: 134,139 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00493766
CAS-Nummer: 18591-78-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR