2,3-Dihydro-1-benzofuran-5-ol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-5-hydroxybenzo[b]furan, 2,3-Dihydrobenzofuran-5-ol

OR42003-1G OR42003-250MG
APOSOR42003-1GEA 294 EUR
APOSOR42003-1G APOSOR42003-250MG
2,3-Dihydro-1-benzofuran-5-ol 97%
2,3-Dihydro-1-benzofuran-5-ol
Formel: C₈H₈O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09029838
CAS-Nummer: 40492-52-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR