Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: Bis(2-cyanoethyl) diisopropylphosphoramidite, Bis(2-cyanoethoxy)(diisopropylamino)phosphine, Bis(2-cyanoethoxy)-N,N-diisopropylaminophosphine

OR4200T-1G OR4200T-5G
APOSOR4200T-1GEA 29 EUR
APOSOR4200T-1G APOSOR4200T-5G
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite 96%
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite

A useful phosphorylating reagent.

Formel: C₁₂H₂₂N₃O₂P
Lagertemperatur: Gefrierschrank
MDL: MFCD00797597
CAS-Nummer: 102690-88-0
UN: 3278
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR