Methyl-3-(chlorosulphonyl)-4-methoxybenzoate 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Methyl 3-(chlorosulphonyl)-4-methoxybenzoate

OR42015-1G
APOSOR42015-1GEA 194 EUR
APOSOR42015-1G
Methyl-3-(chlorosulphonyl)-4-methoxybenzoate 95%
Methyl-3-(chlorosulphonyl)-4-methoxybenzoate
Formel: C₉H₉ClO₅S
Molecular Weight: 264,69 g/mol
Schmelzpunkt: 126…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03407355
CAS-Nummer: 192323-12-9
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR