4-Chinolinol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Quinolin-4-ol, 4-Hydroxyquinoline

OR4202-100G
APOSOR4202-100GEA 79.9 EUR
APOSOR4202-100G
4-Chinolinol 97%
4-Chinolinol
Formel: C₉H₇NO
Molecular Weight: 145.16 g/mol
Schmelzpunkt: 200…206 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006777
CAS-Nummer: 611-36-9
EINECS: 210-268-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR