3-Acetyl-6,8-dichlorocoumarin

Lieferant: Apollo Scientific

Synonyme: 3-Acetyl-6,8-dichloro-2H-chromen-2-one, 3-Acetyl-6,8-dichloro--2-oxo-2H-chromene

OR42021-1G
APOSOR42021-1GEA 116 EUR
APOSOR42021-1G
3-Acetyl-6,8-dichlorocoumarin
3-Acetyl-6,8-dichlorocoumarin
Formel: C₁₁H₆Cl₂O₃
Molecular Weight: 257,07 g/mol
Schmelzpunkt: 172…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00087010
CAS-Nummer: 2199-91-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR