2-Chloro-3-iodopyridin-4-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-2-chloro-3-iodopyridine

OR42029-1G
APOSOR42029-1GEA 21.9 EUR
APOSOR42029-1G
2-Chloro-3-iodopyridin-4-amine
2-Chloro-3-iodopyridin-4-amine
Formel: C₅H₄ClIN₂
Molecular Weight: 254,455 g/mol
Schmelzpunkt: 116…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09763660
CAS-Nummer: 909036-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR