6-Bromo-4-chloro-8-methylquinoline

Lieferant: Apollo Scientific
Achtung

OR42031-1G
APOSOR42031-1GEA 433 EUR
APOSOR42031-1G
6-Bromo-4-chloro-8-methylquinoline
6-Bromo-4-chloro-8-methylquinoline
Formel: C₁₀H₇BrClN
Molecular Weight: 256,526 g/mol
Schmelzpunkt: 153…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12192753
CAS-Nummer: 1086062-90-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR