7-Bromo-4-chloro-8-methylquinoline

Lieferant: Apollo Scientific

OR42032-1G
APOSOR42032-1GEA 119 EUR
APOSOR42032-1G
7-Bromo-4-chloro-8-methylquinoline
7-Bromo-4-chloro-8-methylquinoline
Formel: C₁₀H₇BrClN
Molecular Weight: 256,526 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12674986
CAS-Nummer: 1189106-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR