7-Bromo-4-chloro-6-methylquinoline
Lieferant: Apollo Scientific
OR42036-1G
APOSOR42036-1GEA
509
EUR
APOSOR42036-1G
7-Bromo-4-chloro-6-methylquinoline
7-Bromo-4-chloro-6-methylquinoline
Formel:
C₁₀H₇BrClN Molecular Weight: 256,526 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12674996 CAS-Nummer: 1189105-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...