4-Amino-7-bromo-6-methylquinoline

Lieferant: Apollo Scientific

Synonyme: 7-Bromo-6-methylquinolin-4-amine

OR42037-1G
APOSOR42037-1GEA 607 EUR
APOSOR42037-1G
4-Amino-7-bromo-6-methylquinoline
4-Amino-7-bromo-6-methylquinoline
Formel: C₁₀H₉BrN₂
Molecular Weight: 237,096 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12674999
CAS-Nummer: 1189106-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR