6-Bromo-4-chloro-2,8-dimethylquinoline

Lieferant: Apollo Scientific

OR42038-1G
APOSOR42038-1GEA 433 EUR
APOSOR42038-1G
6-Bromo-4-chloro-2,8-dimethylquinoline
6-Bromo-4-chloro-2,8-dimethylquinoline
Formel: C₁₁H₉BrClN
Molecular Weight: 270,553 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12114952
CAS-Nummer: 1153002-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR