7-Bromo-4-chloro-2,8-dimethylquinoline

Lieferant: Apollo Scientific

OR42039-1G
APOSOR42039-1GEA 282 EUR
APOSOR42039-1G
7-Bromo-4-chloro-2,8-dimethylquinoline
7-Bromo-4-chloro-2,8-dimethylquinoline
Formel: C₁₁H₉BrClN
Lagertemperatur: Raumtemperatur
MDL: MFCD12675042
CAS-Nummer: 1189106-62-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR