2-Bromothiazole-4-carboxamide 97%

Lieferant: Apollo Scientific
Achtung

OR42063-1G
APOSOR42063-1GEA 43.5 EUR
APOSOR42063-1G
2-Bromothiazole-4-carboxamide 97%
2-Bromothiazole-4-carboxamide
Formel: C₄H₃BrN₂OS
Lagertemperatur: Kühlschrank
MDL: MFCD11870082
CAS-Nummer: 848501-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR