2-Indanylacetic acid 99%

Lieferant: Apollo Scientific

Synonyme: 2-(2,3-Dihydro-1H-inden-2-yl)acetic acid

OR42111-1G OR42111-5G
APOSOR42111-1GEA 130 EUR
APOSOR42111-1G APOSOR42111-5G
2-Indanylacetic acid 99%
2-Indanylacetic acid
Formel: C₁₁H₁₂O₂
Schmelzpunkt: 91…93 °C
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082653
CAS-Nummer: 37868-26-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR