7-Bromo-6-methyl-1H-indazole
Lieferant: Apollo Scientific
|
Achtung
|
OR42116-250MG
APOSOR42116-250MGEA
197
EUR
APOSOR42116-250MG
7-Bromo-6-methyl-1H-indazole
7-Bromo-6-methyl-1H-indazole
Formel:
C₈H₇BrN₂ Molecular Weight: 211,059 g/mol Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD17168273 CAS-Nummer: 1257535-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...