5-(Bromomethyl)isoquinoline hydrobromide

Lieferant: Apollo Scientific
Achtung

OR42118-250MG
APOSOR42118-250MGEA 105 EUR
APOSOR42118-250MG
5-(Bromomethyl)isoquinoline hydrobromide
5-(Bromomethyl)isoquinoline hydrobromide
Formel: C₁₀H₈BrN·BrH
Molecular Weight: 302,993 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11976354
CAS-Nummer: 586373-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR