Methyl-4-bromo-1H-pyrazole-3-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-bromo-1H-pyrazole-3-carboxylate

OR42123-1G
APOSOR42123-1GEA 21.9 EUR
APOSOR42123-1G
Methyl-4-bromo-1H-pyrazole-3-carboxylate
Methyl-4-bromo-1H-pyrazole-3-carboxylate
Formel: C₅H₅BrN₂O₂
Molecular Weight: 205,0094 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00466326
CAS-Nummer: 81190-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR