3-Amino-4-methoxybenzonitril 97%

Lieferant: Apollo Scientific
Achtung

OR42124-1G
APOSOR42124-1GEA 71.2 EUR
APOSOR42124-1G
3-Amino-4-methoxybenzonitril 97%
3-Amino-4-methoxybenzonitril
Formel: C₈H₈N₂O
Molecular Weight: 148,1600 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09745274
CAS-Nummer: 60979-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR