4-(2-Amino-1,3-thiazol-4-yl)-N,N-dimethylbenzenesulphonamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-[4-(N,N-dimethylsulphamoyl)phenyl]-1,3-thiazole, 4-(2-Aminothiazol-4-yl)-N,N-dimethylbenzenesulphonamide, 2-Amino-4-{4-[(dimethylamino)sulphonyl]phenyl}-1,3-thiazole

OR42125-250MG
APOSOR42125-250MGEA 242 EUR
APOSOR42125-250MG
4-(2-Amino-1,3-thiazol-4-yl)-N,N-dimethylbenzenesulphonamide
4-(2-Amino-1,3-thiazol-4-yl)-N,N-dimethylbenzenesulphonamide
Formel: C₁₁H₁₃N₃O₂S₂
Molecular Weight: 283.37 g/mol
Schmelzpunkt: 245 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03444373
CAS-Nummer: 610275-82-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR