4-Pyrimidinemethanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Hydroxymethyl)pyrimidine
OR42131-1G
APOSOR42131-1GEA
155
EUR
APOSOR42131-1G
4-Pyrimidinemethanol
4-Pyrimidinemethanol
Formel:
C₅H₆N₂O Molecular Weight: 110,12 g/mol Schmelzpunkt: 75…78 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD03789625 CAS-Nummer: 33581-98-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...