3-Amino-5-brom-2-methoxypyridin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-methoxypyridin-3-amine

OR42139-100G
APOSOR42139-100GEA 183 EUR
APOSOR42139-100G
3-Amino-5-brom-2-methoxypyridin 98%
3-Amino-5-bromo-2-methoxypyridine
Formel: C₆H₇BrN₂O
Molecular Weight: 203.04 g/mol
Schmelzpunkt: 57…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368887
CAS-Nummer: 884495-39-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR